Anti-NFE2L2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137550-50
ABCAAB137550-50EA 973 EUR
ABCAAB137550-50
Anti-NFE2L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFE2L2 (nuclear factor, erythroid 2-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR