Anti-AMPD2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137598-50
ABCAAB137598-50EA 877 EUR
ABCAAB137598-50
Anti-AMPD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AMPD2 (adenosine monophosphate deaminase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR