Anti-RGL2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137600-50
ABCAAB137600-50EA 837 EUR
ABCAAB137600-50
Anti-RGL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RGL2 (ral guanine nucleotide dissociation stimulator-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR