Anti-HAX1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137613-100EA 796 EUR
ABCAAB137613-100
Anti-HAX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HAX1 (HCLS1 associated protein X-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human