Anti-IFRD1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137633-50
ABCAAB137633-50EA 877 EUR
ABCAAB137633-50
Anti-IFRD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IFRD1 (interferon-related developmental regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR