Anti-CYP2R1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137634-50EA 902 EUR
ABCAAB137634-50
Anti-CYP2R1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP2R1 (cytochrome P450, family 2, subfamily R, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR