Anti-KLF5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137676-100EA 883 EUR
ABCAAB137676-100
Anti-KLF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLF5 (Kruppel-like factor 5 (intestinal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR