Anti-NAGLU Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137685-100
ABCAAB137685-100EA 882 EUR
ABCAAB137685-100
Anti-NAGLU Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAGLU (N-acetylglucosaminidase, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR