Anti-FGF21 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137715-100EA 859 EUR
ABCAAB137715-100
Anti-FGF21 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGF21 (fibroblast growth factor 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human