Anti-TH Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137721-100EA 862 EUR
ABCAAB137721-100
Anti-TH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR