Anti-FGG Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137728-100
ABCAAB137728-100EA 882 EUR
ABCAAB137728-100
Anti-FGG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGG (fibrinogen gamma chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR