Anti-P cadherin Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137729-100EA 840 EUR
ABCAAB137729-100
Anti-P cadherin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: P cadherin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human