Anti-ANXA5 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB137823-100
ABCAAB137823-100EA 882 EUR
ABCAAB137823-100
Anti-ANXA5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANXA5 (annexin A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR