Anti-TBX5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137833-100EA 854 EUR
ABCAAB137833-100
Anti-TBX5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TBX5 (T-box 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR