Anti-Seladin 1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB137845-100EA 859 EUR
ABCAAB137845-100
Anti-Seladin 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Seladin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse