Anti-RAB20 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB137943-100EA 796 EUR
ABCAAB137943-100
Anti-RAB20 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB20 (RAB20, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR