Anti-SRSF7 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB138022-100
ABCAAB138022-100EA 882 EUR
ABCAAB138022-100
Anti-SRSF7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SRSF7 (serine/arginine-rich splicing factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR