Anti-STK11 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB138386-100
ABCAAB138386-100EA 882 EUR
ABCAAB138386-100
Anti-STK11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STK11 (serine/threonine kinase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR