Anti-PDLIM5 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB138402-100EA 840 EUR
ABCAAB138402-100
Anti-PDLIM5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDLIM5 (PDZ and LIM domain 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human