Anti-pro Caspase 9 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB138412-100EA 835 EUR
ABCAAB138412-100
Anti-pro Caspase 9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: pro Caspase 9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR