Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 3G12]

Lieferant: Abcam
ABCAAB138463-50EA 807 EUR
ABCAAB138463-50
Anti-14-3-3 eta Mouse Monoclonal Antibody [clone: 3G12]
Antikörper
Type: Primary
Antigen: 14-3-3 eta (14-3-3 eta)
Clonality: monoclonal
Clone: 3G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR