Anti-RAB22A Rabbit Monoclonal Antibody

Lieferant: Abcam
AB138505-100
ABCAAB138505-100EA 876 EUR
ABCAAB138505-100
Anti-RAB22A Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RAB22A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR