Anti-SREBF1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB138663-100EA 840 EUR
ABCAAB138663-100
Anti-SREBF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human