Anti-NPRB Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB139188-100EA 866 EUR
ABCAAB139188-100
Anti-NPRB Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NPRB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen