Anti-PCDH7 Mouse Monoclonal Antibody [clone: 2G6]

Lieferant: Abcam
ABCAAB139274-100EA 869 EUR
ABCAAB139274-100
Anti-PCDH7 Mouse Monoclonal Antibody [clone: 2G6]
Antikörper
Type: Primary
Antigen: PCDH7 (protocadherin 7)
Clonality: monoclonal
Clone: 2G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR