Anti-TICAM1 Mouse Monoclonal Antibody [clone: 1G7]
Lieferant: Abcam
ABCAAB139281-100EA 811 EUR
ABCAAB139281-100
Anti-TICAM1 Mouse Monoclonal Antibody [clone: 1G7]
Type: Primary
Antigen: TICAM1 (toll-like receptor adaptor molecule 1)
Clonality: monoclonal
Clone: 1G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen