Anti-GBAS Mouse Monoclonal Antibody [clone: 1B8]
Lieferant: Abcam
ABCAAB139357-100EA 755 EUR
ABCAAB139357-100
Anti-GBAS Mouse Monoclonal Antibody [clone: 1B8]
Type: Primary
Antigen: GBAS (glioblastoma amplified sequence)
Clonality: monoclonal
Clone: 1B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen