Anti-THY1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5E10]
Lieferant: Abcam
ABCAAB139364-50EA 413 EUR
ABCAAB139364-50
Anti-THY1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5E10]
Antikörper
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: 50000000000
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen