Anti-MHC Class II Rat Monoclonal Antibody [clone: M5/114]

Lieferant: Abcam
AB139365-100
ABCAAB139365-100EA 900 EUR
ABCAAB139365-100
Anti-MHC Class II Rat Monoclonal Antibody [clone: M5/114]
Antikörper
Type: Primary
Antigen: MHC Class II
Clonality: monoclonal
Clone: M5/114
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR