Anti-CCR1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB139399-50EA 840 EUR
ABCAAB139399-50
Anti-CCR1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCR1 (chemokine (C-C motif) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR