Anti-AKR1B10 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB139685-100EA 858 EUR
ABCAAB139685-100
Anti-AKR1B10 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1B10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen