Anti-LMX1A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB139726-100EA 866 EUR
ABCAAB139726-100
Anti-LMX1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMX1A (LIM homeobox transcription factor 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat