Anti-CBX4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB139815-100EA 819 EUR
ABCAAB139815-100
Anti-CBX4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBX4 (chromobox homolog 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR