Anti-SELK Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB139949-100EA 835 EUR
ABCAAB139949-100
Anti-SELK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SELK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR