Anti-PMPCA Rabbit Polyclonal Antibody

Lieferant: Abcam
AB140171-100
ABCAAB140171-100EA 837 EUR
ABCAAB140171-100
Anti-PMPCA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PMPCA (peptidase (mitochondrial processing) alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR