Anti-FOXRED2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140187-100EA 797 EUR
ABCAAB140187-100
Anti-FOXRED2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOXRED2 (FAD-dependent oxidoreductase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR