Anti-RILP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140188-100EA 859 EUR
ABCAAB140188-100
Anti-RILP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RILP (Rab interacting lysosomal protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR