Anti-PSMD7 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140428-100EA 835 EUR
ABCAAB140428-100
Anti-PSMD7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSMD7 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR