Anti-NEXN Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140435-100EA 840 EUR
ABCAAB140435-100
Anti-NEXN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEXN (nexilin (F actin binding protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR