Anti-HTR1E Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140506-50EA 797 EUR
ABCAAB140506-50
Anti-HTR1E Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR1E (5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR