Anti-HTR2A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB140524-50EA 840 EUR
ABCAAB140524-50
Anti-HTR2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR2A (5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human