Anti-DHRS7 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB140596-100EA 823 EUR
ABCAAB140596-100
Anti-DHRS7 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DHRS7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen