Anti-KHSRP Rabbit Monoclonal Antibody

Lieferant: Abcam
ABCAAB140648-100EA 860 EUR
ABCAAB140648-100
Anti-KHSRP Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KHSRP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR