Anti-HAS2 Mouse Monoclonal Antibody [clone: 4E7]

Lieferant: Abcam
ABCAAB140671-100EA 900 EUR
ABCAAB140671-100
Anti-HAS2 Mouse Monoclonal Antibody [clone: 4E7]
Antikörper
Type: Primary
Antigen: HAS2 (hyaluronan synthase 2)
Clonality: monoclonal
Clone: 40000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR