Anti-CALCRL Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB140773-50EA 840 EUR
ABCAAB140773-50
Anti-CALCRL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CALCRL (calcitonin receptor-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human