Anti-SLC7A5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140844-50EA 840 EUR
ABCAAB140844-50
Anti-SLC7A5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC7A5 (solute carrier family 7 (amino acid transporter light chain, L system), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR