Anti-MC3R Rabbit Polyclonal Antibody

Lieferant: Abcam
AB140864-50
ABCAAB140864-50EA 877 EUR
ABCAAB140864-50
Anti-MC3R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MC3R (melanocortin 3 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR