Anti-SSTR1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB140945-50EA 840 EUR
ABCAAB140945-50
Anti-SSTR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SSTR1 (somatostatin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR