Anti-STEAP4 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB140951-50
ABCAAB140951-50EA 882 EUR
ABCAAB140951-50
Anti-STEAP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STEAP4 (STEAP family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR