Anti-DMRT1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB140977-50
ABCAAB140977-50EA 882 EUR
ABCAAB140977-50
Anti-DMRT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DMRT1 (doublesex and mab-3 related transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR